APO414279338 Apollo Scientific Ltd

3-Methylsydnone

  • MDL number MFCD25948959
  • Cas Number : 6939-12-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-Methyl-1,2,3-oxadiazol-3-ium-5-olate
Purity 95
Storage temp. Ambient temperature
Physical Form Liquid-Oil
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=55751

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=55751
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart