APO414279389 Apollo Scientific Ltd

6-Chloro-2-methoxypyridine-3-carboxylic acid

  • MDL number MFCD11847277
  • Cas Number : 65515-33-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 170-172
Synonym 6-Chloro-2-methoxynicotinic acid
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/6-chloro-2-methoxypyridine-3-carboxylic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/6-chloro-2-methoxypyridine-3-carboxylic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart