APO414279477 Apollo Scientific Ltd

3-Chloroquinoxaline-2-carboxylic acid

  • MDL number MFCD09864191
  • Cas Number : 20254-76-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/3-chloroquinoxaline-2-carboxylic-acid

Safety Information

signalword TBC
pcodes TBC
MSDS https://store.apolloscientific.co.uk/product/3-chloroquinoxaline-2-carboxylic-acid
pictograms TBC
hstatements TBC
Availability details for
Quantity
  • Availability
  • Add to Cart