APO414279578 Apollo Scientific Ltd

2-Bromo-4-chloro-1-nitrobenzene

  • MDL number MFCD08437614
  • Cas Number : 63860-31-1
  • Molecular Weight : 236.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-bromo-4-chloro-1-nitrobenzene
Inchi Code 1S/C6H3BrClNO2/c7-5-3-4(8)1-2-6(5)9(10)11/h1-3H
Inchi Key VFMAPIFSXMBTQP-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. Ambient Temperature
Physical Form Solid-Low Melt
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-bromo-4-chloro-1-nitrobenzene

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-bromo-4-chloro-1-nitrobenzene
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart