APO414279581 Apollo Scientific Ltd

1,2,3,6-Tetrahydropyridine-4-boronic acid, pinacol ester, N-CBZ protected

  • MDL number MFCD11521562
  • Cas Number : 286961-15-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Benzyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3,6-dihydro-2H-pyridine-1-carboxylate
Purity 97
Storage temp. TBC
Shipping Temperature TBC
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=51796

Safety Information

MSDS http://www.apolloscientific.co.uk/display_item.php?id=51796
Availability details for
Quantity
  • Availability
  • Add to Cart