APO414279636 Apollo Scientific Ltd

2-Acetyl-5-chloro-2H-indazole

  • MDL number MFCD16619588
  • Cas Number : 98083-44-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=53989

Safety Information

signalword Warning
pcodes P280;P305+P351+P338
MSDS http://www.apolloscientific.co.uk/display_item.php?id=53989
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart