APO414279657 Apollo Scientific Ltd

4-Methyl-3-oxo-3,4-dihydro-2H-1,4-benzoxazine-7-carboxylic acid

  • MDL number MFCD11553038
  • Cas Number : 674304-28-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/4-methyl-3-oxo-34-dihydro-2h-14-benzoxazine-7-carboxylic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-methyl-3-oxo-34-dihydro-2h-14-benzoxazine-7-carboxylic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart