APO414279784 Apollo Scientific Ltd

4,8-Dichloro[1]benzofuro[3,2-d]pyrimidine

  • MDL number MFCD00763947
  • Cas Number : 294874-71-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 147-149
Storage temp. Ambient temperature
Physical Form Solid-Crystals
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=53216

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=53216
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart