APO414279870 Apollo Scientific Ltd

3-Oxo-2,3-dihydro-1H-isoindole-4-carboxylic acid

  • MDL number MFCD11506114
  • Cas Number : 935269-27-5
  • Molecular Weight : 177.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-oxo-4-isoindolinecarboxylic acid
Inchi Code 1S/C9H7NO3/c11-8-7-5(4-10-8)2-1-3-6(7)9(12)13/h1-3H,4H2,(H,10,11)(H,12,13)
Inchi Key SONYZPKBPBFLSS-UHFFFAOYSA-N

Description

Synonym 3-Oxo-4-isoindolinecarboxylic acid
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-oxo-23-dihydro-1h-isoindole-4-carboxylic-acid

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/3-oxo-23-dihydro-1h-isoindole-4-carboxylic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart