APO414279923 Apollo Scientific Ltd

2,4-Dichloro-1H-indole-3-carbaldehyde

  • MDL number MFCD11520132
  • Cas Number : 28035-67-8
  • Molecular Weight : 214.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/24-dichloro-1h-indole-3-carbaldehyde

Safety Information

signalword Warning
pcodes P280;P301+P312;P304+P340
MSDS https://store.apolloscientific.co.uk/product/24-dichloro-1h-indole-3-carbaldehyde
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart