APO414279968 Apollo Scientific Ltd

6-Chloro-1H-indazole-3-carboxaldehyde

  • MDL number MFCD07781580
  • Cas Number : 885521-37-9
  • Molecular Weight : 180.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-chloro-1H-indazole-3-carbaldehyde
Inchi Code 1S/C8H5ClN2O/c9-5-1-2-6-7(3-5)10-11-8(6)4-12/h1-4H,(H,10,11)
Inchi Key YQELBDVPJNKBAP-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/6-chloro-1h-indazole-3-carboxaldehyde

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/6-chloro-1h-indazole-3-carboxaldehyde
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart