APO414280034 Apollo Scientific Ltd

5-Bromo-2-chloroisonicotinonitrile

  • MDL number MFCD16622809
  • Cas Number : 1211515-17-1
  • Molecular Weight : 217.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 79-80
Iupac Name 5-bromo-2-chloroisonicotinonitrile
Inchi Code 1S/C6H2BrClN2/c7-5-3-10-6(8)1-4(5)2-9/h1,3H
Inchi Key IQRASQKEIRAODK-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 79-80
Product Link https://store.apolloscientific.co.uk/product/5-bromo-2-chloroisonicotinonitrile

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/5-bromo-2-chloroisonicotinonitrile
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart