APO414280096 Apollo Scientific Ltd

Methyl 5-bromo-2-methylbenzoate

  • MDL number MFCD09839555
  • Cas Number : 79669-50-4
  • Molecular Weight : 229.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Methyl 5-bromo-o-toluate, 4-Bromo-2-(methoxycarbonyl)toluene
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/methyl-5-bromo-2-methylbenzoate

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-5-bromo-2-methylbenzoate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart