APO414280105 Apollo Scientific Ltd

(2-Chloroethoxy)acetic acid

  • MDL number MFCD17014832
  • Cas Number : 14869-41-1
  • Molecular Weight : 138.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 26-33
Synonym 2-(2-Chloroethoxy)ethanoic acid, 5-Chloro-3-oxapentanoic acid
Purity 97
Density 1.33
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin GB
Boiling point 130/3.5mm
Product Link https://store.apolloscientific.co.uk/product/2-chloroethoxyacetic-acid

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-chloroethoxyacetic-acid
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart