APO414280232 Apollo Scientific Ltd

(Isoquinolin-5-yl)methanamine hydrochloride

  • MDL number MFCD10696103
  • Cas Number : 1001906-56-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/isoquinolin-5-ylmethanamine-hydrochloride

Safety Information

signalword Warning
pcodes P280;P301+P312;P302+P352
MSDS https://store.apolloscientific.co.uk/product/isoquinolin-5-ylmethanamine-hydrochloride
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart