APO414280318 Apollo Scientific Ltd

2-Amino-6-(trifluoromethyl)quinazoline

  • MDL number MFCD00835296
  • Cas Number : 190273-94-0
  • Molecular Weight : 213.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 184-186
Iupac Name 6-(trifluoromethyl)-2-quinazolinamine
Inchi Code 1S/C9H6F3N3/c10-9(11,12)6-1-2-7-5(3-6)4-14-8(13)15-7/h1-4H,(H2,13,14,15)
Inchi Key DFOFJSOWTYTIDK-UHFFFAOYSA-N

Description

Synonym 6-(Trifluoromethyl)quinazolin-2-amine, 2-Amino-6-(trifluoromethyl)-1,3-benzodiazine
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 184-186
Product Link https://store.apolloscientific.co.uk/product/2-amino-6-trifluoromethylquinazoline

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/2-amino-6-trifluoromethylquinazoline
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart