APO414280319 Apollo Scientific Ltd

8-(Trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylic acid monohydrochloride monohydrate

  • MDL number MFCD30184633
  • Cas Number : 1018828-72-2
  • Molecular Weight : 230.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 232-234(dec.)
Iupac Name 8-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylic acid
Inchi Code 1S/C9H5F3N2O2/c10-9(11,12)5-2-1-3-14-4-6(8(15)16)13-7(5)14/h1-4H,(H,15,16)
Inchi Key QRXSLIVFFDRFEB-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 232-234(dec.)
Product Link https://store.apolloscientific.co.uk/product/8-trifluoromethylimidazo12-apyridine-2-carboxylic-acid-monohydrochloride-monohydrate

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/8-trifluoromethylimidazo12-apyridine-2-carboxylic-acid-monohydrochloride-monohydrate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart