APO414280351 Apollo Scientific Ltd

4-Bromo-1-methyl-1H-1,2,3-triazole

  • MDL number MFCD10696344
  • Cas Number : 13273-53-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 98-101
Purity 99
Flash Point >110
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/4-bromo-1-methyl-1h-123-triazole

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-bromo-1-methyl-1h-123-triazole
pictograms GHS07
hstatements H302;H319;H335;H412
Availability details for
Quantity
  • Availability
  • Add to Cart