APO414280361 Apollo Scientific Ltd

3-Amino-5-bromoisoquinoline

  • MDL number MFCD15526591
  • Cas Number : 1192815-01-2
  • Molecular Weight : 223.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-bromo-3-isoquinolinylamine
Inchi Code 1S/C9H7BrN2/c10-8-3-1-2-6-5-12-9(11)4-7(6)8/h1-5H,(H2,11,12)
Inchi Key VZWSVFLRMXYQLA-UHFFFAOYSA-N

Description

Synonym 5-Bromoisoquinolin-3-amine, 3-Amino-5-bromo-2-azanaphthalene
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-amino-5-bromoisoquinoline

Safety Information

signalword Warning
pcodes P260;P280
MSDS https://store.apolloscientific.co.uk/product/3-amino-5-bromoisoquinoline
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart