APO414280394 Apollo Scientific Ltd

5-Hydroxyisoindolin-1-one

  • MDL number MFCD10000830
  • Cas Number : 252061-66-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2,3-Dihydro-5-hydroxy-1H-isoindol-1-one, 2,3-Dihydro-5-hydroxy-1-oxo-1H-isoindole
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/5-hydroxyisoindolin-1-one

Safety Information

signalword Warning
pcodes P280;P271;P260
MSDS https://store.apolloscientific.co.uk/product/5-hydroxyisoindolin-1-one
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart