APO414280484 Apollo Scientific Ltd

2-Acetylbenzonitrile

  • MDL number MFCD00272571
  • Cas Number : 91054-33-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 46-49
Synonym 2-Cyano-acetophenone
Purity 97
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 148/12mm
Product Link https://store.apolloscientific.co.uk/product/2-acetylbenzonitrile

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/2-acetylbenzonitrile
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart