APO414280501 Apollo Scientific Ltd

[(Trimethylsilyl)ethynyl]boronic acid, pinacol ester

  • MDL number MFCD13182130
  • Cas Number : 159087-46-4
  • Molecular Weight : 224.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name trimethyl[(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)ethynyl]silane
Inchi Code 1S/C11H21BO2Si/c1-10(2)11(3,4)14-12(13-10)8-9-15(5,6)7/h1-7H3
Inchi Key WIAMOTQUYWWPCK-UHFFFAOYSA-N

Description

Synonym [(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)ethynyl]trimethylsilane, 4,4,5,5-Tetramethyl-2-[(trimethylsilyl)ethynyl]-1,3,2-dioxaborolane
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/trimethylsilylethynylboronic-acid-pinacol-ester

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/trimethylsilylethynylboronic-acid-pinacol-ester
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart