APO414280592 Apollo Scientific Ltd

3-Chloro-4-(trifluoromethyl)-1H-indazole

  • MDL number MFCD22563699
  • Cas Number : 1388070-12-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=53273

Safety Information

signalword Warning
pcodes P280;P301+P312;P305+P351+P338
MSDS http://www.apolloscientific.co.uk/display_item.php?id=53273
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart