APO414280593 Apollo Scientific Ltd

5-tert-Butyl-1H-imidazole-4-carbaldehyde

  • MDL number MFCD14583325
  • Cas Number : 714273-83-3
  • Molecular Weight : 152.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 122-124
Iupac Name 5-tert-butyl-1H-imidazole-4-carbaldehyde
Inchi Code 1S/C8H12N2O/c1-8(2,3)7-6(4-11)9-5-10-7/h4-5H,1-3H3,(H,9,10)
Inchi Key FSTXHTKEZGSDNN-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 122-124
Product Link https://store.apolloscientific.co.uk/product/5-tert-butyl-1h-imidazole-4-carbaldehyde

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-tert-butyl-1h-imidazole-4-carbaldehyde
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart