APO414280645 Apollo Scientific Ltd

5-Chloro-2-(chloromethyl)pyridine

  • MDL number MFCD10697588
  • Cas Number : 10177-24-9
  • Molecular Weight : 162.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-chloro-2-(chloromethyl)pyridine
Inchi Code 1S/C6H5Cl2N/c7-3-6-2-1-5(8)4-9-6/h1-2,4H,3H2
Inchi Key JTTKXEIHKDSIRC-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Liquid-Oil
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-chloro-2-chloromethylpyridine

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-chloro-2-chloromethylpyridine
pictograms GHS05;GHS07
hstatements H302;H314
Availability details for
Quantity
  • Availability
  • Add to Cart