APO414280725 Apollo Scientific Ltd

6-Bromo-1H-pyrazolo[3,4-b]pyridine

  • MDL number MFCD13178164
  • Cas Number : 934560-92-6
  • Molecular Weight : 198.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 6-Bromo-7-azaindazole
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/6-bromo-1h-pyrazolo34-bpyridine

Safety Information

signalword Warning
pcodes P271;P280
MSDS https://store.apolloscientific.co.uk/product/6-bromo-1h-pyrazolo34-bpyridine
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart