APO414280769 Apollo Scientific Ltd

2,6-Diazaspiro[3.4]octane, N2-BOC protected

  • MDL number MFCD08234737
  • Cas Number : 885270-84-8
  • Molecular Weight : 212.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl 2,6-diazaspiro[3.4]octane-2-carboxylate
Inchi Code 1S/C11H20N2O2/c1-10(2,3)15-9(14)13-7-11(8-13)4-5-12-6-11/h12H,4-8H2,1-3H3
Inchi Key UJIOQJJFPYXAEM-UHFFFAOYSA-N

Description

Synonym tert-Butyl 2,6-diazaspiro[3.4]octane-2-carboxylate, 2-(tert-Butoxycarbonyl)-2,6-diazaspiro[3.4]octane
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/26-diazaspiro34octane-n2-boc-protected

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/26-diazaspiro34octane-n2-boc-protected
pictograms GHS07
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart