APO414280938 Apollo Scientific Ltd

4-Bromo-1H-pyrazolo[3,4-c]pyridin-3-amine

  • MDL number MFCD13177853
  • Cas Number : 870244-35-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/4-bromo-1h-pyrazolo34-cpyridin-3-amine

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-bromo-1h-pyrazolo34-cpyridin-3-amine
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart