APO414281017 Apollo Scientific Ltd

2,5-Diazaspiro[3.5]nonane, N2-BOC protected

  • MDL number MFCD14581200
  • Cas Number : 1246034-93-4
  • Molecular Weight : 226.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym tert-Butyl 2,5-diazaspiro[3.5]nonane-2-carboxylate, 2-(tert-Butoxycarbonyl)-2,5-diazaspiro[3.5]nonane
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/25-diazaspiro35nonane-n2-boc-protected

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/25-diazaspiro35nonane-n2-boc-protected
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart