APO414281028 Apollo Scientific Ltd

3-Nitrobenzhydrazide

  • MDL number MFCD00017059
  • Cas Number : 618-94-0
  • Molecular Weight : 181.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-nitrobenzohydrazide
Inchi Code 1S/C7H7N3O3/c8-9-7(11)5-2-1-3-6(4-5)10(12)13/h1-4H,8H2,(H,9,11)
Inchi Key NQEWXLVDAVTOHM-UHFFFAOYSA-N

Description

Synonym 3-Nitrobenzohydrazide
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-nitrobenzhydrazide

Safety Information

signalword Warning
pcodes P280;P301+P312;P362+P364
MSDS https://store.apolloscientific.co.uk/product/3-nitrobenzhydrazide
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart