APO414281042 Apollo Scientific Ltd

2-Bromoimidazo[1,2-a]pyridine

  • MDL number MFCD09834778
  • Cas Number : 112581-95-0
  • Molecular Weight : 197.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-bromoimidazo12-apyridine

Safety Information

signalword Warning
pcodes P280;P261
MSDS https://store.apolloscientific.co.uk/product/2-bromoimidazo12-apyridine
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart