APO414281079 Apollo Scientific Ltd

2-Hydrazino-5-nitropyridine

  • MDL number MFCD00118332
  • Cas Number : 6343-98-2
  • Molecular Weight : 154.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 209-210
Synonym (5-Nitropyridin-2-yl)hydrazine
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-hydrazino-5-nitropyridine

Safety Information

signalword Warning
pcodes P261;P280;P312
MSDS https://store.apolloscientific.co.uk/product/2-hydrazino-5-nitropyridine
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart