APO414281080 Apollo Scientific Ltd

Ethyl 2-methyl-3-nitrobenzoate

  • MDL number MFCD07359723
  • Cas Number : 59382-60-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 34-36
Synonym 2-(Ethoxycarbonyl)-6-nitrotoluene, Ethyl 3-nitro-o-toluate, 3-(Ethoxycarbonyl)-2-methylnitrobenzene
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=54050

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=54050
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart