APO414281081 Apollo Scientific Ltd

Ethyl [(tert-butoxycarbonyl)amino]acetate

  • MDL number MFCD05663720
  • Cas Number : 14719-37-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Ethyl N-(tert-butoxycarbonyl)glycinate
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=54297

Safety Information

signalword Warning
pcodes P280;P260;P312
MSDS http://www.apolloscientific.co.uk/display_item.php?id=54297
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart