APO414281100 Apollo Scientific Ltd

2,6-Difluoro-3-nitrotoluene

  • MDL number MFCD11519030
  • Cas Number : 79562-49-5
  • Molecular Weight : 173.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 35-38
Synonym 1,3-Difluoro-2-methyl-4-nitrobenzene
Purity 97
Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/26-difluoro-3-nitrotoluene

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/26-difluoro-3-nitrotoluene
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart