APO414281111 Apollo Scientific Ltd

6,7-Dihydro-5H-quinoline-8-one

  • MDL number MFCD03839916
  • Cas Number : 56826-69-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 102-104
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/67-dihydro-5h-quinoline-8-one

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/67-dihydro-5h-quinoline-8-one
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart