APO414281120 Apollo Scientific Ltd

2,4-Dichlorothieno[3,2-d]pyrimidine

  • MDL number MFCD08448158
  • Cas Number : 16234-14-3
  • Molecular Weight : 205.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2,4-dichlorothieno[3,2-d]pyrimidine
Inchi Code 1S/C6H2Cl2N2S/c7-5-4-3(1-2-11-4)9-6(8)10-5/h1-2H
Inchi Key AQECFYPZMBRCIA-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/24-dichlorothieno32-dpyrimidine

Safety Information

signalword Warning
pcodes P280;P301+P312;P261
MSDS https://store.apolloscientific.co.uk/product/24-dichlorothieno32-dpyrimidine
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart