APO414281154 Apollo Scientific Ltd

Ethyl 5-nitro-1-benzothiophene-2-carboxylate

  • MDL number MFCD03791163
  • Cas Number : 25785-09-5
  • Molecular Weight : 251.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/ethyl-5-nitro-1-benzothiophene-2-carboxylate

Safety Information

MSDS https://store.apolloscientific.co.uk/product/ethyl-5-nitro-1-benzothiophene-2-carboxylate
Availability details for
Quantity
  • Availability
  • Add to Cart