APO414281156 Apollo Scientific Ltd

Ethyl (2-methyl-1,3-thiazol-4-yl)acetate

  • MDL number MFCD00052519
  • Cas Number : 37128-24-8
  • Molecular Weight : 185.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/ethyl-2-methyl-13-thiazol-4-ylacetate

Safety Information

MSDS https://store.apolloscientific.co.uk/product/ethyl-2-methyl-13-thiazol-4-ylacetate
Availability details for
Quantity
  • Availability
  • Add to Cart