APO414281157 Apollo Scientific Ltd

2-Methyl-3-nitrobenzhydrazide

  • MDL number MFCD03422927
  • Cas Number : 869942-83-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Methyl-3-nitrobenzohydrazide
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/2-methyl-3-nitrobenzhydrazide

Safety Information

signalword Warning
pcodes P280;P301+P312;P305+P351+P338
MSDS https://store.apolloscientific.co.uk/product/2-methyl-3-nitrobenzhydrazide
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart