APO414281164 Apollo Scientific Ltd

5-Chloroquinazolin-2-amine

  • MDL number MFCD00974335
  • Cas Number : 190273-70-2
  • Molecular Weight : 179.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 249-252
Iupac Name 5-chloro-2-quinazolinamine
Inchi Code 1S/C8H6ClN3/c9-6-2-1-3-7-5(6)4-11-8(10)12-7/h1-4H,(H2,10,11,12)
Inchi Key NUJGYORYWLHYGS-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 249-252
Product Link https://store.apolloscientific.co.uk/product/5-chloroquinazolin-2-amine

Safety Information

signalword Warning
pcodes P280;P305+P351+P338
MSDS https://store.apolloscientific.co.uk/product/5-chloroquinazolin-2-amine
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart