APO414281202 Apollo Scientific Ltd

3-Bromo-9-methyl-9H-carbazole

  • MDL number MFCD00276495
  • Cas Number : 91828-08-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 76-79
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/3-bromo-9-methyl-9h-carbazole

Safety Information

signalword Warning
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-bromo-9-methyl-9h-carbazole
pictograms GHS07
hstatements H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart