APO414281336 Apollo Scientific Ltd

3-Phenylbicyclo[1.1.1]pentane-1-carboxylic acid

  • MDL number MFCD00101889
  • Cas Number : 83249-04-1
  • Molecular Weight : 188.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 176-177
Synonym 1-Carboxy-3-phenylbicyclo[1.1.1]pentane
Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-phenylbicyclo111pentane-1-carboxylic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-phenylbicyclo111pentane-1-carboxylic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart