APO414281377 Apollo Scientific Ltd

7-Nitro-2H-1,4-benzoxazin-3(4H)-one

  • MDL number MFCD00090355
  • Cas Number : 81721-86-0
  • Molecular Weight : 194.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 7-nitro-2H-1,4-benzoxazin-3(4H)-one
Inchi Code 1S/C8H6N2O4/c11-8-4-14-7-3-5(10(12)13)1-2-6(7)9-8/h1-3H,4H2,(H,9,11)
Inchi Key YVGHCFMAEHXPBH-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/7-nitro-2h-14-benzoxazin-34h-one

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/7-nitro-2h-14-benzoxazin-34h-one
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart