APO414281380 Apollo Scientific Ltd

6-Nitro-2,3-dihydro-1H-isoindol-1-one

  • MDL number MFCD03093931
  • Cas Number : 110568-64-4
  • Molecular Weight : 178.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-nitro-1-isoindolinone
Inchi Code 1S/C8H6N2O3/c11-8-7-3-6(10(12)13)2-1-5(7)4-9-8/h1-3H,4H2,(H,9,11)
Inchi Key QKFIHDGZIPOWKP-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/6-nitro-23-dihydro-1h-isoindol-1-one

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/6-nitro-23-dihydro-1h-isoindol-1-one
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart