APO414281452 Apollo Scientific Ltd

4-Bromophenylboronic acid pinacol ester

  • MDL number MFCD11110553
  • Cas Number : 68716-49-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 71
Synonym 2-(4-Bromophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Purity 98
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/4-bromophenylboronic-acid-pinacol-ester

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/4-bromophenylboronic-acid-pinacol-ester
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart