APO414281484 Apollo Scientific Ltd

Ethyl 3-fluoropyridine-2-carboxylate

  • MDL number MFCD11616884
  • Cas Number : 1187732-69-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 74-76
Purity 95
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/ethyl-3-fluoropyridine-2-carboxylate

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/ethyl-3-fluoropyridine-2-carboxylate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart