APO414281520 Apollo Scientific Ltd

Ethyl 5-bromo-1-benzo[b]thiophene-2-carboxylate

  • MDL number MFCD12756246
  • Cas Number : 13771-68-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=53888

Safety Information

MSDS http://www.apolloscientific.co.uk/display_item.php?id=53888
Availability details for
Quantity
  • Availability
  • Add to Cart