APO414281545 Apollo Scientific Ltd

2,3-Dihydro-1H-isoindol-1-one

  • MDL number MFCD03085939
  • Cas Number : 480-91-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 148-153
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 203-204/18mm
Product Link http://www.apolloscientific.co.uk/display_item.php?id=55251

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=55251
pictograms GHS07
hstatements H319
Availability details for
Quantity
  • Availability
  • Add to Cart