APO414281590 Apollo Scientific Ltd

3-Phenyloxetane-3-carboxylic acid

  • MDL number MFCD20659492
  • Cas Number : 114012-42-9
  • Molecular Weight : 178.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-phenyl-3-oxetanecarboxylic acid
Inchi Code 1S/C10H10O3/c11-9(12)10(6-13-7-10)8-4-2-1-3-5-8/h1-5H,6-7H2,(H,11,12)
Inchi Key WOMJGOIQQFWNKL-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. Store at 2-8
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-phenyloxetane-3-carboxylic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-phenyloxetane-3-carboxylic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart